Learn More
Kingfisher Biotech Inc Canine CD40 Ligand (CD40LG) Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0941D025
Encompass
The Canine CD40 Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine CD40 Ligand applications are for cell culture, ELISA standard, and Western Blot Control. The Canine CD40 Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Canine CD40 Ligand Specifications: (Molecular Weight: 15.8 kDa) (Amino Acid Sequence: MQKGDQDPRI AAHVISEASS NPASVLRWAP KGYYTISSNL VSLENGKQLA VKRQGLYYVY AQVTFCSNRA ASSQAPFVAS LCLHSPSGTE RVLLRAASSR GSSKPCGQQS IHLGGVFELH PGASVFVNVT DPSQVSHGTG FTSFGLLKL (149)) (Gene ID: 403468). For research use only. Made in the USA
Catalog No. 50-241-3851