Learn More
ACROBiosystems ACROBiosystems Canine CD73 / NT5E Protein, His Tag
Catalog No. 502106818
Encompass_Preferred
Recombinant Protein;100UG;Canine CD73, His Tag (CD3-C52H5) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 549 (Accession # XP_038540011.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.
Catalog No. 50-210-6818 Supplier ACROBiosystems Supplier No. CD3C52H5100UG
Recombinant Protein;100UG;Canine CD73, His Tag (CD3-C52H5) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 549 (Accession # XP_038540011.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.