Learn More
ACROBiosystems ACROBiosystems Canine CD73 / NT5E Protein, His Tag
Supplier:  ACROBiosystems CD3C52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Canine CD73, His Tag (CD3-C52H5) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 549 (Accession # XP_038540011.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.
Catalog No. 50-210-6817