Learn More
Abcam Canine Complement C3 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1576971X96TESTS
Encompass_Preferred
Canine Complement C3 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine Complement C3 in Dog in Plasma, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2282375