Learn More
American Research Products Inc Canine connectiVe tissue growt
Catalog No. 5000329285
Encompass
Canine connectiVe tissue growth factor CTGF ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29285 Supplier American Research Products Inc Supplier No. CSBE13386C
Canine connectiVe tissue growth factor CTGF ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich