Learn More
Kingfisher Biotech Inc Canine CXCL10 (IP-10) Recombinant Protein (Yeast, Endotoxin - Free) - 500 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0754D500
Encompass
The Canine CXCL10 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine CXCL10 applications are for cell culture, ELISA standard, and Western Blot Control. The Canine CXCL10 yeast-derived recombinant protein can be purchased in multiple sizes. Canine CXCL10 Specifications: (Molecular Weight: 8.6 kDa) (Amino Acid Sequence: IPLSRTIRCT CIKISDQPVN LRSLEKIEMI PASPSCPHVE IIATMKKSGE KRCLNPESKT IKSLVKAISK KRSRRSP (77)) (Gene ID: 478432). For research use only. Made in the USA
Catalog No. 50-241-3227