Learn More
Sino Biological Anti-BCA-1/CXCL13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 70057T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine BCA-1 / CXCL13 (Catalog#70057-D07E; XP_851619.3; Val23-Asn110). BCA-1 / CXCL13 specific IgG was purified by Canine BCA-1 / CXCL13 affinity chromatography.
Catalog No. 50-241-1631