Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine/Mouse Cystatin C ELISA Kit

Catalog No. EC2RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EC2RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EC2RBX10 Supplier Invitrogen™ Supplier No. EC2RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Canine Cystatin C quantitates canine Cystatin C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant canine Cystatin C.

Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21460
Assay Range 10-2500 pg/mL
Assay Sensitivity 10 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse, Canine
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227;
Gene ID (Entrez) 13010
Gene Symbol Cst3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Cystatin C
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.