Learn More
Raybiotech Inc C-Series Canine Cytokine Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AACCYT12
Encompass_Preferred
Detects 40 Canine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Cystatin C, Decorin, EGFR, ErbB3, Erythropoietin, FGF-7 (KGF), Galectin-3, GASP-1 (WFIKKNRP), GM-CSF, GRO alpha (CXCL1), HGF, HGFR, IFN-gamma, IGFBP-2, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 Ra (IL-1 F3), IL-10, IL-12 p40, IL-13, IL-17A, IL-2, IL-21, IL-6, IL-8 (CXCL8), MCP-1 (CCL2), MIP-1 beta (CCL4), MMP-8, NCAM-1 (CD56), Nope, PDGF-BB, RAGE, RANTES (CCL5), Resistin, SCF, TGF alpha, TNF alpha, TNF RI (TNFRSF1A), Trappin-2, VEGF-A
Catalog No. 50-194-8449