Learn More
Raybiotech Inc G-Series Canine Cytokine Array 1 Kit.
Catalog No. 501950339
Encompass_Preferred
Semi-quantitative measurement of 10 Canine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IL-2, IL-6, IL-8 (CXCL8), IL-10, GM-CSF, MCP-1 (CCL2), RAGE, SCF, TNF alpha, VEGF-A
Catalog No. 50-195-0339 Supplier Raybiotech Inc Supplier No. GSCCYT12
Semi-quantitative measurement of 10 Canine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IL-2, IL-6, IL-8 (CXCL8), IL-10, GM-CSF, MCP-1 (CCL2), RAGE, SCF, TNF alpha, VEGF-A