Learn More
Raybiotech Inc G-Series Canine Cytokine Array 2 Kit.
Catalog No. 501950340
Encompass_Preferred
Semi-quantitative measurement of 10 Canine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Erythropoietin, FGF-7 (KGF), HGF, HGFR, IFN-gamma, IL-1 beta (IL-1 F2), IL-12 p40, IL-17A, MIP-1 beta (CCL4), TNF RI (TNFRSF1A)
Catalog No. 50-195-0340 Supplier Raybiotech Inc Supplier No. GSCCYT21
Semi-quantitative measurement of 10 Canine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Erythropoietin, FGF-7 (KGF), HGF, HGFR, IFN-gamma, IL-1 beta (IL-1 F2), IL-12 p40, IL-17A, MIP-1 beta (CCL4), TNF RI (TNFRSF1A)