Learn More
Raybiotech Inc G-Series Canine Cytokine Array 3 Kit.
Catalog No. 501950344
Encompass_Preferred
Detects 10 Canine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Cystatin C, Decorin, EGFR, IL-1 alpha, IL-1ra, IL-13, IL-21, RANTES (CCL5), Resistin, Trappin-2 (Elafin)
Catalog No. 50-195-0344 Supplier Raybiotech Inc Supplier No. GSCCYT41
Detects 10 Canine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Cystatin C, Decorin, EGFR, IL-1 alpha, IL-1ra, IL-13, IL-21, RANTES (CCL5), Resistin, Trappin-2 (Elafin)