Learn More
Raybiotech Inc G-Series Canine Cytokine Array 40 Kit.
Catalog No. 501950337
Encompass_Preferred
A combination of 4 non-overlapping arrays to semi-quantitatively measure 40 Canine cytokines. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Cystatin C, Decorin, EGFR, ErbB3, Erythropoietin, FGF-7, Galectin-3, GASP-1, GM-CSF, GRO alpha, HGF, HGFR, IFN-gamma, IGFBP-2, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p40, IL-13, IL-17A, IL-2, IL-21, IL-6, IL-8, MCP-1, MIP-1 beta, MMP-8, NCAM-1, Nope, PDGF-BB, RAGE, RANTES, Resistin, SCF, TGF alpha, TNF alpha, TNF RI, Trappin-2, VEGF-A
Catalog No. 50-195-0337 Supplier Raybiotech Inc Supplier No. GSCCAA402
A combination of 4 non-overlapping arrays to semi-quantitatively measure 40 Canine cytokines. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Cystatin C, Decorin, EGFR, ErbB3, Erythropoietin, FGF-7, Galectin-3, GASP-1, GM-CSF, GRO alpha, HGF, HGFR, IFN-gamma, IGFBP-2, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p40, IL-13, IL-17A, IL-2, IL-21, IL-6, IL-8, MCP-1, MIP-1 beta, MMP-8, NCAM-1, Nope, PDGF-BB, RAGE, RANTES, Resistin, SCF, TGF alpha, TNF alpha, TNF RI, Trappin-2, VEGF-A