Learn More
Raybiotech Inc Quantibody Canine Cytokine Antibody Array 40 Kit.
Supplier:  Raybiotech Inc QACCAA401
Encompass_Preferred
A combination of 4 non-overlapping arrays to quantitatively measure 40 Canine cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Cystatin C, Decorin, EGFR, ErbB3, Erythropoietin, FGF-7, Galectin-3, GASP-1, GM-CSF, GRO alpha, HGF, HGFR, IFN-gamma, IGFBP-2, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p40, IL-13, IL-17A, IL-2, IL-21, IL-6, IL-8, MCP-1, MIP-1 beta, MMP-8, NCAM-1, Nope, PDGF-BB, RAGE, RANTES, Resistin, SCF, TGF alpha, TNF alpha, TNF RI, Trappin-2, VEGF-A
Catalog No. 50-195-0782