Learn More
American Research Products Inc Canine Dihydrotestosterone DH
Catalog No. 5000326490
Encompass
Canine Dihydrotestosterone DHT ELISA Kit from CUSABIO Detection Range 40 pg/mL-8000 pg/mL Sensitivity 20 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26490 Supplier American Research Products Inc Supplier No. CSBE07881C
Canine Dihydrotestosterone DHT ELISA Kit from CUSABIO Detection Range 40 pg/mL-8000 pg/mL Sensitivity 20 pg/mL Sample serum plasma tissue homogenates Method Competitive