Learn More
Vmrd Inc CANINE DISTEMPER VIRUS 0.1MG
SDP
Supplier:  Vmrd Inc 1C42H11
Encompass
Specific for Canine Distemper Virus envelope component; neutralizes Rockborn, Snyder Hill and Onderstepoort Strains. IgG1 isotype.
Catalog No. NC0637665