Learn More
ATCC Canine distemper virus; Strain: Snyder Hill
Catalog No. 502382118
Encompass
Canine distemper virus strain Snyder Hill was derived from ATCC VR-526 by adaptation to growth in Vero cells (ATCC CCL-81). ATCC VR-526, deposited by L.E. Carmichael, was isolated from the brain of a dog with clinical distemper in September 1950. Because it was propagated in animal tissue, ATCC will no longer offer that product.
Catalog No. 50-238-2118 Supplier ATCC Supplier No. VR1587
Canine distemper virus strain Snyder Hill was derived from ATCC VR-526 by adaptation to growth in Vero cells (ATCC CCL-81). ATCC VR-526, deposited by L.E. Carmichael, was isolated from the brain of a dog with clinical distemper in September 1950. Because it was propagated in animal tissue, ATCC will no longer offer that product.