Learn More
Kingfisher Biotech Inc Canine Fas Ligand Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1088D025
Encompass
The Canine Fas Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine Fas Ligand applications are for cell culture, ELISA standard, and Western Blot Control. Canine Fas Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Canine Fas Ligand Specifications: (Molecular Weight: 17.3 kDa) (Amino Acid Sequence: QIGQPNPPSE KRELRKVAHL TGKPNSRSIP LEWEDTYGIA LVSGVKYKKG GLVINDTGLY FVYSKVYFRG QSCNNKPLNH KVYMRNSKYP QDLMLMEGKI MNYCTTGQMW ARSSYLGAVF NLTSADHLYV NVSELSLVSF EESKTFFGLY KL (152)) (Gene ID: 442968). For research use only. Made in the USA
Catalog No. 50-253-3697