Learn More
Sino Biological Recombinant Canine CD64/FCGR1 Protein (His Tag), HPLC-verified
Catalog No. 502992773
Encompass_Preferred
A DNA sequence encoding the Canine FCGR1 (NP_001002976.1) (Val11-Pro288) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2773 Supplier Sino Biological Supplier No. 70269D08H100
A DNA sequence encoding the Canine FCGR1 (NP_001002976.1) (Val11-Pro288) was expressed with a polyhistidine tag at the C-terminus.