Learn More
Sino Biological Anti-FGF1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 70039T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine aFGF / FGF1 (Catalog#70039-DNAE; XP_849274.1; Phe16-Asp155). aFGF / FGF1 specific IgG was purified by Canine aFGF / FGF1 affinity chromatography.
Catalog No. 50-255-2903