Learn More
American Research Products Inc Canine follicle stimulating ho
Catalog No. 5000326289
Encompass
Canine follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 0 35 mIU/mL-140 mIU/mL Sensitivity 0 35 mIU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26289 Supplier American Research Products Inc Supplier No. CSBE06868C
Canine follicle-stimulating hormone FSH ELISA Kit from CUSABIO Detection Range 0 35 mIU/mL-140 mIU/mL Sensitivity 0 35 mIU/mL Sample serum plasma tissue homogenates Method Competitive