Learn More
Sino Biological Anti-FOLR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 70115RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine FOLR1 / Folate Receptor 1 (Catalog#70115-D08H; XP_851993.1; Met1-Met228). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2409