Learn More
American Research Products Inc Canine Free ThyroXine FT4 ELI
Catalog No. 5000326049
Encompass
Canine Free ThyroXine FT4 ELISA Kit from CUSABIO Detection Range 7 5 ng/mL-120 ng/mL Sensitivity 7 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26049 Supplier American Research Products Inc Supplier No. CSBE13196C
Canine Free ThyroXine FT4 ELISA Kit from CUSABIO Detection Range 7 5 ng/mL-120 ng/mL Sensitivity 7 5 ng/mL Sample serum plasma tissue homogenates Method Competitive