Learn More
American Research Products Inc Canine Free Tri iodothyronine
Catalog No. 5000329998
Encompass
Canine Free Tri-iodothyronine (Free-T3) ELISA Kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma Method Competitive
Catalog No. 50-003-29998 Supplier American Research Products Inc Supplier No. CSBE13195C
Canine Free Tri-iodothyronine (Free-T3) ELISA Kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma Method Competitive