Learn More
Raybiotech Inc Canine Galectin-3 ELISA Kit
Supplier:  Raybiotech Inc ELCGALECTIN31
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Canine Galectin-3 (LGALS3)
Catalog No. 50-194-8882