Learn More
Raybiotech Inc Canine GM-CSF ELISA Kit
Supplier:  Raybiotech Inc ELCGMCSF1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.13 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Canine GM-CSF.
Catalog No. 50-194-8883