Learn More
Raybiotech Inc Canine GRO alpha/CXCL1 ELISA Kit
Supplier:  Raybiotech Inc ELCCXCL11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 5.4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Canine GRO alpha.
Catalog No. 50-194-8878