Learn More
Sino Biological Recombinant Canine GUCY2C/Guanylyl cyclase C Protein (ECD, His Tag)
Supplier:  Sino Biological 70264D08H100
Encompass_Preferred
A DNA sequence encoding the canine GUCY2C (F1PAG3) (Met1-Gln430) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6549