Learn More
Sino Biological Recombinant Anti-TIM-1/HAVCR1 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 70001R201H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine TIM-1 / KIM-1 / HACVR ( Catalog#70001-D07H; XP_854888.1; Tyr21-Ser160), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2040