Learn More
Sino Biological Canine HAVCR1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 70001R201100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine TIM-1 / KIM-1 / HACVR ( Catalog#70001-D07H; XP_854888.1; Tyr21-Ser160).
Catalog No. 50-237-1931