Learn More
Immunology Consultants Canine Heartworm (D. immitis) Antigen, 1.0 mg
SDP
Supplier:  Immunology Consultants RA40HRT
Encompass_Preferred
Washed, adult female worms were disrupted, homogenized and protein was extracted. Total protein measurement was determined by Bio-Rad Protein Assay using BGG as a standard. This product shows multiple bands as determined by SDS-PAGE gel. It is suitable for use as a control in ELISA, blotting or other immunoassays. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-9041