Learn More
Sino Biological Recombinant Anti-HGF Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 70063R105100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine HGF/Hepatocyte Growth Factor ( Catalog#70063-DNAH; NP_001002964.1; Met1-Ser730).
Catalog No. 50-255-2901