Learn More
Kingfisher Biotech Inc Canine IFN beta 1 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1026D025
Encompass
The Canine IFN beta 1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IFN beta 1 applications are for cell culture, ELISA standard, and Western Blot Control. Canine IFN beta 1 yeast-derived recombinant protein can be purchased in multiple sizes. Canine IFN beta 1 Specifications: (Molecular Weight: 20.0 kDa) (Amino Acid Sequence: MSNDLLRSQL SSSSLECQEL LLQLNGTTEY CLKDRINFEI PEEIEKSRQF QKEDIILITH EMFQKIFDIF RRNISRTGWN ETTVENLLVK LHWQKEHLEI ILEDVKEKEN FTWDNRTLLH LKKYYLRIVQ YLKAKEYSIC AWTIVQAEIC RNFFFLNILT DYLQN (165)) (Gene ID: 481558). For research use only. Made in the USA
Catalog No. 50-241-3675