Learn More
Kingfisher Biotech Inc Canine IFN gamma Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0271D025
Encompass
The Canine IFN gamma yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IFN gamma applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IFN gamma yeast-derived recombinant protein can be purchased in multiple sizes. Canine IFN gamma Specifications: (Molecular Weight: 16.7 kDa) (Amino Acid Sequence: QAMFFKEIEN LKEYFNASNP DVSDGGSLFV DILKKWREES DKTIIQSQIV SFYLKLFDNF KDNQIIQRSM DTIKEDMLGK FLNSSTSKRE DFLKLIQIPV NDLQVQRKAI NELIKVMNDL SPRSNLRKRK RSQNLFRGRR ASK (143)) (Gene ID: 403801). For research use only. Made in the USA
Catalog No. 50-241-4017