Learn More
Abcam Canine IgE ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1577001X96TESTS
Encompass_Preferred
Canine IgE ELISA Kit ab157700 is used to quantify Canine IgE with a sensitivity of 2.29 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1883377