Learn More
Raybiotech Inc Canine IGFBP-2 ELISA Kit
Supplier:  Raybiotech Inc ELCIGFBP21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.055 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Canine IGFBP-2.
Catalog No. 50-194-8887