Learn More
Kingfisher Biotech Inc Canine IL-1 alpha Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0486D025
Encompass
The Canine IL-1 alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-1 alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-1 alpha yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-1 alpha Specifications: (Molecular Weight: 18.2 kDa) (Amino Acid Sequence: SVAYNFHNNE KYNYIRIIKS QFILNDNLNQ SIVRQTGGNY LMTAALQNLD DAVKFDMGAY TSEDSKLPVT LRISKTRLFV SAQNEDEPVL LKEMPETPKT IRDETNLLFF WERHGSKHYF KSVAQPKLFI ATQERKLVHM ARGQPSITDF RLLETQP (157)) (Gene ID: 403782). For research use only. Made in the USA
Catalog No. 50-241-4032