Learn More
Kingfisher Biotech Inc Canine IL-1 beta Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0085D005
Encompass
The Canine IL-1 beta yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-1 beta applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-1 beta yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-1 beta Specifications: (Molecular Weight: 17.5 kDa) (Amino Acid Sequence: AAMQSVDCKLQDISHKYLVLSNSYELRALHLNGENVNKQVVFHMSFVHGDESNNKIPVVLGIKQKNLYLSCVMKDGKPTLQLEKVDPKVYPKRKMEKRFVFNKIEIKNTVEFESSQYPNWYISTSQVEGMPVFLGNTRGGQDITDFTMEFSS) (Gene ID: 403974). For research use only. Made in the USA
Catalog No. 50-241-4050