Learn More
Kingfisher Biotech Inc Canine IL-1 Receptor Antagonist Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0427D025
Encompass
The Canine IL-1 Receptor Antagonist yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-1 Receptor Antagonist applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-1 Receptor Antagonist yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-1 Receptor Antagonist Specifications: (Molecular Weight: 16.8 kDa) (Amino Acid Sequence: LGKRPCRMQAFRIWDVNQKTFYLRNNQLVAGYLQGSNTKLEEKLDVVPVEPHAVFLGIHGGKLCLACVKSGDETRLQLEAVNITDLSKNKDQDKRFTFILSDSGPTTSFESAACPGWFLCTALEADRPVSLTNRPEEAMMVTKFYFQKE (149)) (Gene ID: 403660). For research use only. Made in the USA
Catalog No. 50-241-3707