Learn More
Raybiotech Inc Canine IL-12 p40 ELISA Kit
Supplier:  Raybiotech Inc ELCIL12P401
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 24 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Canine IL-12 p40.
Catalog No. 50-194-8889