Learn More
Kingfisher Biotech Inc Canine IL-13 Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc KP1368D100
Encompass
The Canine IL-13 polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Canine IL-13 ELISA. The Canine IL-13 antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine IL-13 - None) (Canine IL-13 - Strong) (Caprine IL-13 - None) (Chicken IL-13 - None) (Dolphin IL-13 - Weak) (Equine IL-13 - None) (Human IL-13 - None) (Mouse IL-13 - None) (Ovine IL-13 - None) (Rabbit IL-13 - None) (Swine IL-13 - None) (Zebrafish IL-13 - None). For research use only. Made in the USA
Catalog No. 50-233-2255