Learn More
Kingfisher Biotech Inc Canine IL-13 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0737D025
Encompass
The Canine IL-13 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-13 applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-13 yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-13 Specifications: (Molecular Weight: 12.3 kDa) (Amino Acid Sequence: SPSPVTPSPT LKELIEELVN ITQNQASLCN GSMVWSVNLT AGMYCAALES LINVSDCSAI QRTQRMLKAL CSQKPAAGIS SERSRDTKIE VIQLVKNLLT YVRGVYRHGN FR (112)) (Gene ID: 442990). For research use only. Made in the USA
Catalog No. 50-241-3158