Learn More
Kingfisher Biotech Inc Swine CXCL9 (MIG) ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1370D004
Encompass_Preferred
The Swine CXCL9 (MIG) ELISA contains capture antibody, protein standard, and detection antibody for development of a Swine CXCL9 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CXCL9, CMK, Humig, MIG, SCYB9, crg-10, C-X-C motif chemokine ligand 9. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5432