Learn More
ACROBiosystems ACROBiosystems Canine IL-13 R alpha 2 Protein, His Tag
Catalog No. 502106267
Encompass_Preferred
Recombinant Protein;100UG;Canine IL-13 R alpha 2, His Tag (IL2-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Thr 338 (Accession # Q95LF0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.3 kDa.
Catalog No. 50-210-6267 Supplier ACROBiosystems Supplier No. IL2C52H6100UG
Recombinant Protein;100UG;Canine IL-13 R alpha 2, His Tag (IL2-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Thr 338 (Accession # Q95LF0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.3 kDa.