Learn More
Kingfisher Biotech Inc Canine IL-17A Recombinant Protein (Yeast, Endotoxin - Free) - 500 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0920D500
Encompass
The Canine IL-17A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-17A applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-17A yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-17A Specifications: (Molecular Weight: 14.8 kDa) (Amino Acid Sequence: FPQNPGCRNT EDKNFPQHVK VNLNILNRNT NSRRPSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CVNNEGNINY HMNSVPIQQE ILVLRRESQH CPHSFRLEKM LVAVGCTCVT PIVRHVA (127)) (Gene ID: 481837). For research use only. Made in the USA
Catalog No. 50-241-4004