Learn More
Kingfisher Biotech Inc Canine IL-17A Polyclonal Antibody - Biotinylated (50 ug)
SDP
Supplier:  Kingfisher Biotech Inc KPB0959D050
Encompass
The Canine IL-17A polyclonal antibody is labeled and has been qualified for use in ELISA. It is the detection antibody in the Kingfisher Biotech Canine IL-17A ELISA. The Canine IL-17A antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine IL-17A - None) (Canine IL-17A - Strong) (Equine IL-17A - None) (Human IL-17A - Strong) (Mouse IL-17A - None) (Ovine IL-17A - None) (Rabbit IL-17A - None) (Rat IL-17A - None) (Swine IL-17A - None). For research use only. Made in the USA
Catalog No. 50-233-2501