Learn More
ACROBiosystems ACROBiosystems Canine IL-17A / CTLA-8 Protein, His Tag
Catalog No. 502106394
Encompass_Preferred
Recombinant Protein;1MG;Canine IL-17A, His Tag (ILA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Ala 155 (Accession # NP_001159350.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.0 kDa.
Catalog No. 50-210-6394 Supplier ACROBiosystems Supplier No. ILAC52H71MG
Recombinant Protein;1MG;Canine IL-17A, His Tag (ILA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Ala 155 (Accession # NP_001159350.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.0 kDa.