Learn More
ACROBiosystems ACROBiosystems Canine IL-2 R alpha / CD25 Protein, His Tag
Catalog No. 502106353
Encompass_Preferred
Recombinant Protein;1MG;Canine IL-2 R alpha, His Tag (ILA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 237 (Accession # O62802-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.4 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine IL-2 R alpha, His Tag (Cat. No. ILA-C52H4) at 5 μg/mL (100 μL/well) can bind Human IL-2 N-Fc & C-Avitag with a linear range of 0.1-10 ng/mL (QC tested).
Catalog No. 50-210-6353 Supplier ACROBiosystems Supplier No. ILAC52H41MG
Recombinant Protein;1MG;Canine IL-2 R alpha, His Tag (ILA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 237 (Accession # O62802-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.4 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Canine IL-2 R alpha, His Tag (Cat. No. ILA-C52H4) at 5 μg/mL (100 μL/well) can bind Human IL-2 N-Fc & C-Avitag with a linear range of 0.1-10 ng/mL (QC tested).