Learn More
ACROBiosystems ACROBiosystems Canine IL-2 R beta Protein, His Tag
Catalog No. 502106465
Encompass_Preferred
Recombinant Protein;50UG;Canine IL-2 R beta, His Tag (ILA-C52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 30 - Pro 246 (Accession # F1PGA6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.9 kDa.
Catalog No. 50-210-6465 Supplier ACROBiosystems Supplier No. ILAC52H550UG
Recombinant Protein;50UG;Canine IL-2 R beta, His Tag (ILA-C52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 30 - Pro 246 (Accession # F1PGA6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.9 kDa.