Learn More
Kingfisher Biotech Inc Canine IL-24 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0293D025
Encompass
The Canine IL-24 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-24 applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-24 yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-24 Specifications: (Molecular Weight: 18.2 kDa) (Amino Acid Sequence: QEFRFGPCRV QGVALRELRE AFWTVKDTVQ AKDNITSVRL LRKEVLQDVS DAESCYLIRA LLKFYLNTVF KNYLDEAADV RIRRSFSTLA NNFFVIASKL QPSQEDEMFS ISESARRRFL LFQRAFKQLD IQAAQTKAFG EVDILLTWME KFYEF (155)) (Gene ID: 609213). For research use only. Made in the USA
Catalog No. 50-241-3399